1,2-Diazepan-1-yl(4-nitrophenyl)methanone ≥95%
Supplier: Matrix Scientific
046273-500MG
046273-1G
101874-904EA
1109.23
USD
101874-904
101874-906
1,2-Diazepan-1-yl(4-nitrophenyl)methanone ≥95%
1,2-Diazepan-1-yl(4-nitrophenyl)methanone
Hazard: Irritant
Formula:
C₁₂H₁₅N₃O₃ Melting Pt: 82…86 °C |
MDL Number:
MFCD11840984 CAS Number: 1135283-09-8 |
Specification Test Results
Melting point | 82-86°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...