6-Chloro-5-iodo-2-methyl-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

046269-500MG 046269-1G
101874-894EA 556.92 USD
101874-894 101874-896
6-Chloro-5-iodo-2-methyl-4-pyrimidinol ≥95%
6-Chloro-5-iodo-2-methyl-4-pyrimidinol

Hazard: Irritant

Formula: C₅H₄ClIN₂O
Melting Pt: 255…257 °C
MDL Number: MFCD12025846
CAS Number: 1194-73-6

Order Now

Specification Test Results

Melting point 255-257°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR