1-Benzyl-2-methyl-5-nitro-1H-1,3-benzimidazole ≥95%
Supplier: Matrix Scientific
046268-500MG
046268-1G
101874-890EA
375.38
USD
101874-890
101874-892
1-Benzyl-2-methyl-5-nitro-1H-1,3-benzimidazole ≥95%
1-Benzyl-2-methyl-5-nitro-1H-1,3-benzimidazole
Hazard: Irritant
Formula:
C₁₅H₁₃N₃O₂ Melting Pt: 176…178 °C |
MDL Number:
MFCD08688839 CAS Number: 14624-88-5 |
Specification Test Results
Melting point | 176-178°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...