5,6-Dichloro-2-methyl-4-pyrimidinol ≥95%
Supplier: Matrix Scientific
046262-500MG
046262-1G
101874-866EA
556.92
USD
101874-866
101874-868
5,6-Dichloro-2-methyl-4-pyrimidinol ≥95%
5,6-Dichloro-2-methyl-4-pyrimidinol
Hazard: Irritant
Formula:
C₅H₄Cl₂N₂O Melting Pt: 249…251 °C |
MDL Number:
MFCD12025845 CAS Number: 32265-50-2 |
Specification Test Results
Melting point | 249-251°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...