tert-Butyl-2-(4-nitrobenzoyl)-1,2-diazepane-1-carboxylate ≥95%

Supplier: Matrix Scientific

046257-500MG 046257-1G
101874-852EA 747.69 USD
101874-852 101874-854
tert-Butyl-2-(4-nitrobenzoyl)-1,2-diazepane-1-carboxylate ≥95%
tert-Butyl-2-(4-nitrobenzoyl)-1,2-diazepane-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₃N₃O₅
MW: 349.39 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD11840975
CAS Number: 1135283-01-0

Order Now

Specification Test Results

Melting point 110-112°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR