6-(Dimethylamino)-2-methyl-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

046255-500MG 046255-1G
101874-846EA 375.38 USD
101874-846 101874-848
6-(Dimethylamino)-2-methyl-4-pyrimidinol ≥95%
6-(Dimethylamino)-2-methyl-4-pyrimidinol

Hazard: Irritant

Formula: C₇H₁₁N₃O
Melting Pt: 226…228 °C
MDL Number: MFCD11840974
CAS Number: 83724-17-8

Order Now

Specification Test Results

Melting point 226-228°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR