[4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-yl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
[4'-(1,3-Dioxolan-2-yl)-[1,1'-biphenyl]-4-yl]methanol
046253-500MG
046253-1G
101874-838EA
556.92
USD
101874-838
101874-840
[4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-yl]methanol ≥95%
[4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-yl]methanol
Hazard: Irritant
Formula:
C₁₆H₁₆O₃ MW: 256.3 g/mol Melting Pt: 122…125 °C |
MDL Number:
MFCD06802587 CAS Number: 889954-12-5 |
Specification Test Results
Melting point | 122-125°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...