Methyl-2-chloro-5-fluoro-6-methoxynicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-5-fluoro-6-methoxynicotinate

046252-500MG 046252-1G
101874-834EA 223.08 USD
101874-834 101874-836
Methyl-2-chloro-5-fluoro-6-methoxynicotinate ≥95%
Methyl-2-chloro-5-fluoro-6-methoxynicotinate

Hazard: Irritant

Formula: C₈H₇ClFNO₃
MW: 219.6 g/mol
Melting Pt: 66…68 °C
MDL Number: MFCD12025844
CAS Number: 959616-64-9

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR