2-Chloro-5-fluoro-6-methoxynicotinic acid ≥95%

Supplier: Matrix Scientific

046251-500MG 046251-1G
101874-830EA 375.38 USD
101874-830 101874-832
2-Chloro-5-fluoro-6-methoxynicotinic acid ≥95%
2-Chloro-5-fluoro-6-methoxynicotinic acid

Hazard: Irritant

Formula: C₇H₅ClFNO₃
Melting Pt: 164…166 °C
MDL Number: MFCD12025843
CAS Number: 943025-86-3

Order Now

Specification Test Results

Melting point 164-166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR