2-(2-Fluoro-5-nitrophenyl)-1H-isoindole-1,3(2H)-dione ≥95%

Supplier: Matrix Scientific

046247-500MG 046247-1G
101874-818EA 278.46 USD
101874-818 101874-820
2-(2-Fluoro-5-nitrophenyl)-1H-isoindole-1,3(2H)-dione ≥95%
2-(2-Fluoro-5-nitrophenyl)-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₄H₇FN₂O₄
Melting Pt: 154…156 °C
MDL Number: MFCD01119713
CAS Number: 166658-69-1

Order Now

Specification Test Results

Melting point 154-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR