N-(2-Fluoro-5-nitrophenyl)-N-(methylsulfonyl)methanesulfonamide ≥95%
Supplier: Matrix Scientific
046245-500MG
046245-1G
101874-810EA
278.46
USD
101874-810
101874-812
N-(2-Fluoro-5-nitrophenyl)-N-(methylsulfonyl)methanesulfonamide ≥95%
N-(2-Fluoro-5-nitrophenyl)-N-(methylsulfonyl)methanesulfonamide
Hazard: Irritant
Formula:
C₈H₉FN₂O₆S₂ Melting Pt: 133…135 °C |
MDL Number:
MFCD11840970 CAS Number: 1135282-97-1 |
Specification Test Results
Melting point | 133-135°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...