N-(2-Fluoro-5-nitrophenyl)methanesulfonamide ≥95%

Supplier: Matrix Scientific

046243-500MG 046243-1G
101874-804EA 278.46 USD
101874-804 101874-806
N-(2-Fluoro-5-nitrophenyl)methanesulfonamide ≥95%
N-(2-Fluoro-5-nitrophenyl)methanesulfonamide

Hazard: Irritant

Formula: C₇H₇FN₂O₄S
Melting Pt: 163…165 °C
MDL Number: MFCD01213923
CAS Number: 123343-99-7

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR