N-(2-Fluoro-5-nitrophenyl)acetamide ≥95%

Supplier: Matrix Scientific

046239-500MG 046239-1G 046239-5G
101874-788EA 181.54 USD
101874-788 101874-790 101874-792
N-(2-Fluoro-5-nitrophenyl)acetamide ≥95%
N-(2-Fluoro-5-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₈H₇FN₂O₃
Melting Pt: 180…182 °C
MDL Number: MFCD00661814
CAS Number: 454-07-9

Order Now

Specification Test Results

Melting point 180-182°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR