6-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

046237-500MG 046237-1G
101874-780EA 375.38 USD
101874-780 101874-782
6-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide ≥95%
6-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₁₀H₇F₃N₂OS
MW: 260.24 g/mol
Melting Pt: 183…185 °C
MDL Number: MFCD11615820
CAS Number: 1135282-94-8

Order Now

Specification Test Results

Melting point 183-185°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR