2-Amino-6-chloronicotinic acid ≥95%

Supplier: Matrix Scientific

046233-250MG 046233-1G
101874-764EA 143.08 USD
101874-764 101874-766
2-Amino-6-chloronicotinic acid ≥95%
2-Amino-6-chloronicotinic acid

Hazard: Irritant

Formula: C₆H₅ClN₂O₂
Melting Pt: 195…197 °C (dec.)
MDL Number: MFCD11045328
CAS Number: 58584-92-2

Order Now

Specification Test Results

Melting point 195-197°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR