3-Bromo-6-fluoroimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

046232-500MG 046232-1G
101874-760EA 349.23 USD
101874-760 101874-762
3-Bromo-6-fluoroimidazo[1,2-a]pyridine ≥95%
3-Bromo-6-fluoroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄BrFN₂
MW: 215.200 g/mol
Melting Pt: 141-142°C
MDL Number: MFCD12827770
CAS Number: 1186405-11-7

Order Now

Specification Test Results

Melting point 141-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR