3-tert-Butyl-5-(4-nitrophenyl)-1,2,4-oxadiazole ≥95%

Supplier: Matrix Scientific

046231-500MG 046231-1G 046231-5G
101874-754EA 278.46 USD
101874-754 101874-756 101874-758
3-tert-Butyl-5-(4-nitrophenyl)-1,2,4-oxadiazole ≥95%
3-tert-Butyl-5-(4-nitrophenyl)-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₁₂H₁₃N₃O₃
Melting Pt: 142-144°C
MDL Number: MFCD11840954
CAS Number: 1135282-84-6

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR