3-[4-(4-Methylpiperazino)phenyl]-3-oxopropanenitrile ≥95%

Supplier: Matrix Scientific

046230-500MG 046230-1G
101874-750EA 375.38 USD
101874-750 101874-752
3-[4-(4-Methylpiperazino)phenyl]-3-oxopropanenitrile ≥95%
3-[4-(4-Methylpiperazino)phenyl]-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₄H₁₇N₃O
MW: 243.31 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD12025842
CAS Number: 1135282-83-5

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR