3-Oxo-3-[4-(1H-pyrazol-1-yl)phenyl]propanenitrile ≥95%
Supplier: Matrix Scientific
046223-500MG
046223-1G
101874-726EA
415.38
USD
101874-726
101874-728
3-Oxo-3-[4-(1H-pyrazol-1-yl)phenyl]propanenitrile ≥95%
3-Oxo-3-[4-(1H-pyrazol-1-yl)phenyl]propanenitrile
Hazard: Irritant
Formula:
C₁₂H₉N₃O Melting Pt: 170…172 °C |
MDL Number:
MFCD12025838 CAS Number: 1176541-33-5 |
Specification Test Results
Melting point | 170-172°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...