7-Fluoro-2H-benzo[b][1,4]oxazin-3(4H)-one ≥95%

Supplier: Matrix Scientific

046220-500MG 046220-1G
101874-720EA 192.31 USD
101874-720 101874-722
7-Fluoro-2H-benzo[b][1,4]oxazin-3(4H)-one ≥95%
7-Fluoro-2H-benzo[b][1,4]oxazin-3(4H)-one

Hazard: Irritant

Formula: C₈H₆FNO₂
Melting Pt: 203…205 °C
MDL Number: MFCD07774200
CAS Number: 103361-99-5

Order Now

Specification Test Results

Melting point 203-205°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR