3-Amino-5-fluoro-1H-indazole ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Fluoro-1H-indazol-3-amine
046214-500MG
046214-1G
101874-704EA
361.54
USD
101874-704
101874-706
3-Amino-5-fluoro-1H-indazole ≥95%
3-Amino-5-fluoro-1H-indazole
Hazard: Irritant
Formula:
C₇H₆FN₃ MW: 151.141 g/mol Melting Pt: 165-167°C |
MDL Number:
MFCD03426699 CAS Number: 61272-72-8 |
Specification Test Results
Melting point | 165-167°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...