tert-Butyl-4-[6-chloro-5-(hydroxymethyl)-4-pyrimidinyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

046212-500MG 046212-1G 046212-5G
101874-692EA 500 USD
101874-692 101874-694 101874-696
tert-Butyl-4-[6-chloro-5-(hydroxymethyl)-4-pyrimidinyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-[6-chloro-5-(hydroxymethyl)-4-pyrimidinyl]tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₄H₂₁ClN₄O₃
MW: 328.8 g/mol
Melting Pt: 151…153 °C
MDL Number: MFCD09972226
CAS Number: 1017782-56-7

Order Now

Specification Test Results

Melting point 151-153°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR