5-(tert-Butyl)-3-ethyl-4,6-dioxo-3a,4,6,6a-tetrahydropyrrolo[3,4-c]pyrazole-3,5(1H)-dicarboxylate ≥95%

Supplier: Matrix Scientific

046210-500MG 046210-1G
101874-682EA 375.38 USD
101874-682 101874-684
5-(tert-Butyl)-3-ethyl-4,6-dioxo-3a,4,6,6a-tetrahydropyrrolo[3,4-c]pyrazole-3,5(1H)-dicarboxylate ≥95%
5-(tert-Butyl)-3-ethyl-4,6-dioxo-3a,4,6,6a-tetrahydropyrrolo[3,4-c]pyrazole-3,5(1H)-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₁₇N₃O₆
Melting Pt: 124…125 °C
MDL Number: MFCD09972224
CAS Number: 1171423-05-4

Order Now

Specification Test Results

Melting point 124-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR