1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone ≥95%
Supplier: Matrix Scientific
046207-500MG
046207-1G
101874-678EA
556.92
USD
101874-678
101874-680
1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone ≥95%
1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone
Hazard: Irritant
Formula:
C₁₀H₇BrO₂ Melting Pt: 89…91 °C |
MDL Number:
MFCD09972223 CAS Number: 99661-02-6 |
Specification Test Results
Melting point | 89-91°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...