2'-(Methylsulfinyl)[1,1'-biphenyl]-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
046206-500MG
046206-1G
101874-674EA
375.38
USD
101874-674
101874-676
2'-(Methylsulfinyl)[1,1'-biphenyl]-4-carbaldehyde ≥95%
2'-(Methylsulfinyl)[1,1'-biphenyl]-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₂O₂S Melting Pt: 123…124 °C |
MDL Number:
MFCD09972222 CAS Number: 1171155-44-4 |
Specification Test Results
Melting point | 123-124°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...