(4'-Nitro[1,1'-biphenyl]-4-yl)methanol ≥95%
Supplier: Matrix Scientific
046205-500MG
046205-1G
101874-670EA
556.92
USD
101874-670
101874-672
(4'-Nitro[1,1'-biphenyl]-4-yl)methanol ≥95%
(4'-Nitro[1,1'-biphenyl]-4-yl)methanol
Hazard: Irritant
Formula:
C₁₃H₁₁NO₃ Melting Pt: 165 °C |
MDL Number:
MFCD09972221 CAS Number: 62037-99-4 |
Specification Test Results
Melting point | 165°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...