3-Bromo-1-benzofuran ≥95%
Supplier: Matrix Scientific
046201-500MG
046201-1G
046201-5G
101874-656EA
3474.92
USD
101874-656
101874-658
101874-660
3-Bromo-1-benzofuran ≥95%
3-Bromo-1-benzofuran
Hazard: Irritant/Store in Freezer
Formula:
C₈H₅BrO Melting Pt: 39…40 °C |
MDL Number:
MFCD01029425 CAS Number: 59214-70-9 |
Specification Test Results
Melting point | 39-40°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...