3,5-Dibromo-1-methyl-4-nitro-1H-pyrazole ≥95%
Supplier: Matrix Scientific
046197-500MG
046197-1G
101874-638EA
561.54
USD
101874-638
101874-640
3,5-Dibromo-1-methyl-4-nitro-1H-pyrazole ≥95%
3,5-Dibromo-1-methyl-4-nitro-1H-pyrazole
Hazard: Irritant
Formula:
C₄H₃Br₂N₃O₂ Melting Pt: 153…155 °C |
MDL Number:
MFCD09972218 CAS Number: 155600-99-0 |
Specification Test Results
Melting point | 153-155°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...