Methyl-3'-nitro[1,1'-biphenyl]-4-carboxylate ≥95%
Supplier: Matrix Scientific
046195-500MG
046195-1G
046195-5G
101874-626EA
181.54
USD
101874-626
101874-628
101874-630
Methyl-3'-nitro[1,1'-biphenyl]-4-carboxylate ≥95%
Methyl-3'-nitro[1,1'-biphenyl]-4-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₁NO₄ Melting Pt: 144…146 °C |
MDL Number:
MFCD01846431 CAS Number: 89900-93-6 |
Specification Test Results
Melting point | 144-146°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...