[5-(4-Chlorophenyl)-1,3,4-oxadiazol-2-yl]-N-methylmethanamine ≥95%
Supplier: Matrix Scientific
046194-500MG
046194-1G
101874-622EA
921.54
USD
101874-622
101874-624
[5-(4-Chlorophenyl)-1,3,4-oxadiazol-2-yl]-N-methylmethanamine ≥95%
[5-(4-Chlorophenyl)-1,3,4-oxadiazol-2-yl]-N-methylmethanamine
Hazard: Irritant
Formula:
C₁₀H₁₀ClN₃O MW: 223.66 g/mol Melting Pt: 56…58 °C |
MDL Number:
MFCD09972216 CAS Number: 1017782-52-3 |
Specification Test Results
Melting point | 56-58°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...