Ethyl-2-(2,4-difluoro-5-nitrophenoxy)acetate ≥95%
Supplier: Matrix Scientific
046192-500MG
046192-1G
046192-5G
101874-614EA
556.92
USD
101874-614
101874-616
101874-618
Ethyl-2-(2,4-difluoro-5-nitrophenoxy)acetate ≥95%
Ethyl-2-(2,4-difluoro-5-nitrophenoxy)acetate
Hazard: Irritant
Formula:
C₁₀H₉F₂NO₅ Melting Pt: 47…49 °C |
MDL Number:
MFCD09972214 CAS Number: 179011-36-0 |
Specification Test Results
Melting point | 47-49°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...