N-(6-Chloro-2-phenyl-4-pyrimidinyl)-N-ethylamine ≥95%

Supplier: Matrix Scientific

046191-500MG 046191-1G 046191-5G
101874-608EA 375.38 USD
101874-608 101874-610 101874-612
N-(6-Chloro-2-phenyl-4-pyrimidinyl)-N-ethylamine ≥95%
N-(6-Chloro-2-phenyl-4-pyrimidinyl)-N-ethylamine

Hazard: Irritant

Formula: C₁₂H₁₂ClN₃
Melting Pt: 69…71 °C
MDL Number: MFCD09972213
CAS Number: 26871-14-7

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR