Ethyl-3-[4-(benzyloxy)phenyl]acrylate ≥95%
Supplier: Matrix Scientific
046190-500MG
046190-1G
101874-604EA
400
USD
101874-604
101874-606
Ethyl-3-[4-(benzyloxy)phenyl]acrylate ≥95%
Ethyl-3-[4-(benzyloxy)phenyl]acrylate
Hazard: Irritant
Formula:
C₁₈H₁₈O₃ Melting Pt: 96…97 °C |
MDL Number:
MFCD01570263 CAS Number: 104315-07-3 |
Specification Test Results
Melting point | 96-97°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...