4-Chloro-2-phenyl-6-piperidinopyrimidine ≥95%
Supplier: Matrix Scientific
046188-500MG
046188-1G
046188-5G
101874-594EA
375.38
USD
101874-594
101874-596
101874-598
4-Chloro-2-phenyl-6-piperidinopyrimidine ≥95%
4-Chloro-2-phenyl-6-piperidinopyrimidine
Hazard: Irritant
Formula:
C₁₅H₁₆ClN₃ Melting Pt: 124…126 °C |
MDL Number:
MFCD00754197 CAS Number: 574708-35-3 |
Specification Test Results
Melting point | 124-126°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...