N-(6-Chloro-2-phenyl-4-pyrimidinyl)-N,N-dimethylamine ≥95%
Supplier: Matrix Scientific
046185-500MG
046185-1G
046185-5G
101874-578EA
400
USD
101874-578
101874-580
101874-582
N-(6-Chloro-2-phenyl-4-pyrimidinyl)-N,N-dimethylamine ≥95%
N-(6-Chloro-2-phenyl-4-pyrimidinyl)-N,N-dimethylamine
Hazard: Irritant
Formula:
C₁₂H₁₂ClN₃ Melting Pt: 94…95 °C |
MDL Number:
MFCD00697491 CAS Number: 58514-86-6 |
Specification Test Results
Melting point | 94-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...