3-{4-[(3,4-Difluorobenzyl)oxy]phenyl}acrylic acid ≥95%

Supplier: Matrix Scientific

046183-500MG 046183-1G 046183-5G
101874-568EA 278.46 USD
101874-568 101874-570 101874-572
3-{4-[(3,4-Difluorobenzyl)oxy]phenyl}acrylic acid ≥95%
3-{4-[(3,4-Difluorobenzyl)oxy]phenyl}acrylic acid

Hazard: Irritant

Formula: C₁₆H₁₂F₂O₃
Melting Pt: 230…232 °C
MDL Number: MFCD09972209
CAS Number: 649740-48-7

Order Now

Specification Test Results

Melting point 230-232°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR