4-[(3,4-Difluorobenzyl)oxy]benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
046182-500MG
046182-1G
046182-5G
101874-562EA
181.54
USD
101874-562
101874-564
101874-566
4-[(3,4-Difluorobenzyl)oxy]benzenecarbaldehyde ≥95%
4-[(3,4-Difluorobenzyl)oxy]benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₀F₂O₂ Melting Pt: 88…90 °C |
MDL Number:
MFCD09972208 CAS Number: 1017782-47-6 |
Specification Test Results
Melting point | 88-90°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...