Methyl-4,5-dibromo-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4,5-dibromothiophene-2-carboxylate

046179-1G 046179-5G
101874-550EA 221.54 USD
101874-550 101874-552
Methyl-4,5-dibromo-2-thiophenecarboxylate ≥95%
Methyl-4,5-dibromo-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₆H₄Br₂O₂S
Melting Pt: 80…82 °C
MDL Number: MFCD00831688
CAS Number: 62224-24-2

Order Now

Specification Test Results

Melting point 80-82°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR