2-Hydroxy-5-iodonicotinic acid ≥95%

Supplier: Matrix Scientific

046178-500MG 046178-1G 046178-5G
101874-542EA 110.77 USD
101874-542 101874-544 101874-546
2-Hydroxy-5-iodonicotinic acid ≥95%
2-Hydroxy-5-iodonicotinic acid

Hazard: Irritant

Formula: C₆H₅NO₃
MW: 265.005 g/mol
Melting Pt: 264-266°C
MDL Number: MFCD11100199
CAS Number: 390360-97-1

Order Now

Specification Test Results

Melting point 264-266°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR