Diethyl-2-[{[(benzyloxy)carbonyl]amino}(3-chlorophenyl)methyl]malonate ≥95%
Supplier: Matrix Scientific
046176-500MG
046176-1G
046176-5G
101874-530EA
278.46
USD
101874-530
101874-532
101874-534
Diethyl-2-[{[(benzyloxy)carbonyl]amino}(3-chlorophenyl)methyl]malonate ≥95%
Diethyl-2-[{[(benzyloxy)carbonyl]amino}(3-chlorophenyl)methyl]malonate
Hazard: Irritant
Formula:
C₂₂H₂₄ClNO₆ Melting Pt: 58…61 °C |
MDL Number:
MFCD10758053 CAS Number: 1033463-15-8 |
Specification Test Results
Melting point | 58-61°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...