2-(4-Benzylpiperidino)acetonitrile ≥95%

Supplier: Matrix Scientific

046173-500MG 046173-1G
101874-518EA 278.46 USD
101874-518 101874-520
2-(4-Benzylpiperidino)acetonitrile ≥95%
2-(4-Benzylpiperidino)acetonitrile

Hazard: Irritant

Formula: C₁₄H₁₈N₂
Melting Pt: 54…56 °C
MDL Number: MFCD09152769
CAS Number: 25842-31-3

Order Now

Specification Test Results

Melting point 54-56°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR