1-(4-Iodophenyl)-2,5-dimethylpyrrole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Iodophenyl)-2,5-dimethyl-1H-pyrrole

046172-500MG 046172-1G 046172-5G
101874-512EA 156.92 USD
101874-512 101874-514 101874-516
1-(4-Iodophenyl)-2,5-dimethylpyrrole ≥95%
1-(4-Iodophenyl)-2,5-dimethylpyrrole

Hazard: Irritant

Formula: C₁₂H₁₂IN
Melting Pt: 82-84°C
MDL Number: MFCD02692647
CAS Number: 288608-09-3

Order Now

Specification Test Results

Melting point 82-84°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR