1-(2-Fluoro-4-iodophenyl)-2,5-dimethyl-1H-pyrrole ≥95%
Supplier: Matrix Scientific
046171-500MG
046171-1G
046171-5G
101874-506EA
240
USD
101874-506
101874-508
101874-510
1-(2-Fluoro-4-iodophenyl)-2,5-dimethyl-1H-pyrrole ≥95%
1-(2-Fluoro-4-iodophenyl)-2,5-dimethyl-1H-pyrrole
Hazard: Irritant
Formula:
C₁₂H₁₁FIN Melting Pt: 70…72 °C |
MDL Number:
MFCD09152719 CAS Number: 217314-30-2 |
Specification Test Results
Melting point | 70-72°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...