1-(3-Iodophenyl)-2,5-dimethylpyrrole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(3-Iodophenyl)-2,5-dimethyl-1H-pyrrole

046170-500MG 046170-1G 046170-5G
101874-500EA 106.15 USD
101874-500 101874-502 101874-504
1-(3-Iodophenyl)-2,5-dimethylpyrrole ≥95%
1-(3-Iodophenyl)-2,5-dimethylpyrrole

Hazard: Irritant

Formula: C₁₂H₁₂IN
Melting Pt: 103-106°C
MDL Number: MFCD07429165
CAS Number: 217314-37-9

Order Now

Specification Test Results

Melting point 103-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR