2-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime ≥95%

Supplier: Matrix Scientific

Synonyms: (E)-N-{[2-(1H-Pyrazol-1-yl)phenyl]methylidene}hydroxylamine

046167-500MG 046167-1G
101874-486EA 375.38 USD
101874-486 101874-488
2-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime ≥95%
2-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime

Hazard: Irritant

Formula: C₁₀H₉N₃O
MW: 187.2 g/mol
Melting Pt: 173…176 °C
MDL Number: MFCD09152718
CAS Number: 1017782-44-3

Order Now

Specification Test Results

Melting point 173-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR