4-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[(5-Bromopyrimidin-2-yl)oxy]benzonitrile

046165-500MG 046165-1G 046165-5G
101874-476EA 921.54 USD
101874-476 101874-478 101874-480
4-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbonitrile ≥95%
4-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₁H₆BrN₃O
MW: 276.1 g/mol
Melting Pt: 145…147 °C
MDL Number: MFCD03646339
CAS Number: 887430-98-0

Order Now

Specification Test Results

Melting point 145-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR