4-Bromo-3-fluoro-2-methyl-6-nitroaniline ≥95%

Supplier: Matrix Scientific

046163-500MG 046163-1G
101874-468EA 415.38 USD
101874-468 101874-470
4-Bromo-3-fluoro-2-methyl-6-nitroaniline ≥95%
4-Bromo-2-fluoro-2-methyl-6-nitroaniline

Hazard: Irritant

Formula: C₇H₆BrFN₂O₂
MW: 249.05 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD09152766
CAS Number: 952183-01-6

Order Now

Specification Test Results

Melting point 124-126°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR