Methyl-4-amino-3-methoxybenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-amino-3-methoxybenzoate
046161-1G
046161-5G
101874-460EA
15.38
USD
101874-460
101874-462
Methyl-4-amino-3-methoxybenzoate ≥95%
Methyl 4-amino-3-methoxybenzoate
Hazard: Irritant
Formula:
C₉H₁₁NO₃ MW: 181.1900 g/mol Melting Pt: 128-131°C |
MDL Number:
MFCD00017203 CAS Number: 41608-64-4 |
Specification Test Results
Melting point | 128-131°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...