1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid, 1-(Ethoxycarbonyl)piperidine-4-carboxylic acid

046160-500MG 046160-1G 046160-5G
101874-454EA 21.54 USD
101874-454 101874-456 101874-458
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid ≥95%
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
Melting Pt: 68…70 °C
MDL Number: MFCD07339354
CAS Number: 118133-15-6

Order Now

Specification Test Results

Melting point 68-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR