3-Chloro-1H-pyrrolo[2,3-b]pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Chloro-7-azaindole
046158-500MG
046158-1G
046158-5G
101874-442EA
278.46
USD
101874-442
101874-444
101874-446
3-Chloro-1H-pyrrolo[2,3-b]pyridine ≥95%
3-Chloro-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol Melting Pt: 168…170 °C |
MDL Number:
MFCD08272228 CAS Number: 80235-01-4 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...